siRNA Details
| Gene Symbol: LURAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 990 | ||
| GenBank mRNA | AK057892.1 | - | 2 | 990 | ||
| BX370151.2 | - | 1 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LURAP1| Gene Symbol: LURAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 990 | ||
| GenBank mRNA | AK057892.1 | - | 2 | 990 | ||
| BX370151.2 | - | 1 | 310 | |||