siRNA Details
| Gene Symbol: KIR2DL5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018081.1 | NP_001018091.1 | 9 | 1582 | ||
| GenBank mRNA | AF271607.1 | - | 9 | 1582 | ||
| AF271608.1 | - | 9 | 1582 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIR2DL5B| Gene Symbol: KIR2DL5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018081.1 | NP_001018091.1 | 9 | 1582 | ||
| GenBank mRNA | AF271607.1 | - | 9 | 1582 | ||
| AF271608.1 | - | 9 | 1582 | |||