siRNA Details
| Gene Symbol: Ncald | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024371.1 | NP_001019542.1 | 4 | 2722 | ||
| XM_008765456.1 | XP_008763678.1 | 5 | 2712 | |||
| GenBank mRNA | BC087105.1 | - | 4 | 2722 | 7193019 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncald| Gene Symbol: Ncald | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024371.1 | NP_001019542.1 | 4 | 2722 | ||
| XM_008765456.1 | XP_008763678.1 | 5 | 2712 | |||
| GenBank mRNA | BC087105.1 | - | 4 | 2722 | 7193019 | |