siRNA Details
| Gene Symbol: FBXO48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024680.2 | NP_001019851.1 | 4 | 1012 | ||
| XM_005264407.2 | XP_005264464.1 | 4 | 1014 | |||
| XM_017004437.1 | XP_016859926.1 | 4 | 1027 | |||
| GenBank mRNA | BC089423.1 | - | 4 | 953 | 3103174 | |
| DQ904332.1 | - | 4 | 953 | |||

FBXO48
Genomic Map