siRNA Details
| Gene Symbol: Olfr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011002.2 | NP_035132.2 | 2 | 1107 | ||
| GenBank mRNA | AF102538.1 | - | 1 | 589 | ||
| BC139159.1 | - | 1 | 867 | 8862181 | ||
| BC139161.1 | - | 1 | 867 | 8862183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr59| Gene Symbol: Olfr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011002.2 | NP_035132.2 | 2 | 1107 | ||
| GenBank mRNA | AF102538.1 | - | 1 | 589 | ||
| BC139159.1 | - | 1 | 867 | 8862181 | ||
| BC139161.1 | - | 1 | 867 | 8862183 | ||