siRNA Details
| Gene Symbol: HES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019089.4 | NP_061962.2 | 4 | 3890 | ||
| GenBank mRNA | AK023754.1 | - | 1 | 3075 | ||
| AK091122.1 | - | 4 | 3890 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HES2| Gene Symbol: HES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019089.4 | NP_061962.2 | 4 | 3890 | ||
| GenBank mRNA | AK023754.1 | - | 1 | 3075 | ||
| AK091122.1 | - | 4 | 3890 | |||