siRNA Details
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 6 | 1045 | ||
NM_031921.5 | NP_114127.3 | 8 | 928 | |||
XM_005244806.3 | XP_005244863.1 | 8 | 951 | |||
XM_011542241.2 | XP_011540543.1 | 8 | 951 | |||
XM_011542243.2 | XP_011540545.1 | 7 | 664 | |||
XM_011542244.1 | XP_011540546.1 | 8 | 1095 | |||
GenBank mRNA | AB033099.1 | - | 6 | 1134 | ||
AK024028.1 | - | 6 | 513 | |||
AK127857.1 | - | 1 | 1579 | |||
AK290586.1 | - | 8 | 920 | |||
AL834179.1 | - | 1 | 495 | |||
BC002542.2 | - | 8 | 854 | 3138578 | ||
BC018701.1 | - | 8 | 854 | 3139886 |
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 4 | 1265 | ||
GenBank mRNA | AK091918.1 | - | 4 | 1266 |
