siRNA Details
| Gene Symbol: Gpr158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004761.1 | NP_001004761.1 | 1 | 1122 | ||
| GenBank mRNA | AB093287.1 | - | 1 | 227 | ||
| AK030530.1 | - | 1 | 1118 | |||
| AK083234.1 | - | 1 | 1148 | |||
| AK086256.1 | - | 1 | 1123 | |||

Gpr158
Genomic Map