siRNA Details
| Gene Symbol: Gphb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007013.1 | NP_001007014.1 | 2 | 376 | ||
| GenBank mRNA | AY437504.1 | - | 2 | 376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gphb5| Gene Symbol: Gphb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007013.1 | NP_001007014.1 | 2 | 376 | ||
| GenBank mRNA | AY437504.1 | - | 2 | 376 | ||