siRNA Details
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007797.1 | NP_001007798.1 | 2 | 333 | ||
GenBank mRNA | AY383703.1 | - | 2 | 333 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007797.1 | NP_001007798.1 | 2 | 333 | ||
GenBank mRNA | AY383703.1 | - | 2 | 333 |