siRNA Details
| Gene Symbol: Rxfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008310.1 | NP_001008311.1 | 1 | 455 | ||
| GenBank mRNA | AY633763.1 | - | 1 | 434 | ||
| AY633764.1 | - | 1 | 455 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxfp3| Gene Symbol: Rxfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008310.1 | NP_001008311.1 | 1 | 455 | ||
| GenBank mRNA | AY633763.1 | - | 1 | 434 | ||
| AY633764.1 | - | 1 | 455 | |||
Set Membership: |
|