siRNA Details
| Gene Symbol: Prl7a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008341.1 | NP_001008342.1 | 2 | 242 | ||
| GenBank mRNA | AY741310.1 | - | 2 | 242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl7a4| Gene Symbol: Prl7a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008341.1 | NP_001008342.1 | 2 | 242 | ||
| GenBank mRNA | AY741310.1 | - | 2 | 242 | ||