siRNA Details
| Gene Symbol: Pan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008862.1 | NP_001008862.1 | 8 | 1385 | ||
| XM_006240783.2 | XP_006240845.1 | 9 | 2553 | |||
| XM_006240784.2 | XP_006240846.1 | 9 | 2553 | |||
| XM_006240785.2 | XP_006240847.1 | 9 | 2553 | |||
| XM_006240786.2 | XP_006240848.1 | 9 | 2553 | |||
| XM_006240787.2 | XP_006240849.1 | 9 | 2553 | |||
| XM_006240788.2 | XP_006240850.1 | 9 | 2553 | |||
| XM_006240789.2 | XP_006240851.1 | 9 | 2553 | |||

Pan2
Genomic Map