siRNA Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008863.3 | NP_001008863.2 | 4 | 422 | ||
| XM_008770560.1 | XP_008768782.1 | 7 | 1785 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp54| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008863.3 | NP_001008863.2 | 4 | 422 | ||
| XM_008770560.1 | XP_008768782.1 | 7 | 1785 | |||