siRNA Details
| Gene Symbol: Zbtb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009172.3 | NP_001009172.1 | 2 | 1066 | ||
| XM_006256123.2 | XP_006256185.1 | 2 | 1280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb22| Gene Symbol: Zbtb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009172.3 | NP_001009172.1 | 2 | 1066 | ||
| XM_006256123.2 | XP_006256185.1 | 2 | 1280 | |||