siRNA Details
| Gene Symbol: Tryx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009173.1 | NP_001009173.1 | 3 | 243 | ||
| XM_006236368.2 | XP_006236430.1 | 3 | 814 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tryx5| Gene Symbol: Tryx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009173.1 | NP_001009173.1 | 3 | 243 | ||
| XM_006236368.2 | XP_006236430.1 | 3 | 814 | |||