siRNA Details
| Gene Symbol: Unc13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009573.2 | NP_001009573.2 | 14 | 1249 | ||
| GenBank mRNA | AK220199.1 | - | 7 | 557 | ||
| BC141289.1 | - | 13 | 1161 | 9055901 | ||
| EF127645.1 | - | 13 | 1197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13d| Gene Symbol: Unc13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009573.2 | NP_001009573.2 | 14 | 1249 | ||
| GenBank mRNA | AK220199.1 | - | 7 | 557 | ||
| BC141289.1 | - | 13 | 1161 | 9055901 | ||
| EF127645.1 | - | 13 | 1197 | |||