siRNA Details
| Gene Symbol: ERMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009959.2 | NP_001009959.1 | 4 | 3045 | ||
| NM_001304344.1 | NP_001291273.1 | 4 | 2893 | |||
| NM_001304345.1 | NP_001291274.1 | 4 | 2905 | |||
| NM_001304346.1 | NP_001291275.1 | 3 | 3018 | |||
| NM_020711.2 | NP_065762.1 | 3 | 3078 | |||
| GenBank mRNA | AB033015.1 | - | 3 | 2820 | ||

ERMN
Genomic Map