siRNA Details
| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012667.2 | NP_001012685.2 | 5 | 1106 | ||
| NM_001146117.1 | NP_001139589.1 | 4 | 1047 | |||
| GenBank mRNA | AK131902.1 | - | 4 | 798 | ||
| AK133712.1 | - | 4 | 1051 | |||
| AK140957.1 | - | 4 | 837 | |||
| BC048089.1 | - | 5 | 1082 | 6466443 | ||

Smim19
Genomic Map