siRNA Details
| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017924.3 | NP_060394.1 | 2 | 347 | ||
| XM_017021390.1 | XP_016876879.1 | 2 | 521 | |||
| GenBank mRNA | AF061731.1 | - | 2 | 317 | ||
| AK000678.1 | - | 2 | 272 | |||
| BC009645.1 | - | 2 | 242 | 3458006 | ||
| BC056255.1 | - | 2 | 318 | 4694305 | ||
| BC061637.1 | - | 2 | 282 | 5532778 | ||
| CR457248.1 | - | 1 | 200 | |||
| DA605680.1 | - | 2 | 347 | |||

C14orf119
Genomic Map