siRNA Details
| Gene Symbol: Inip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013577.1 | NP_001013595.1 | 5 | 1032 | ||
| GenBank mRNA | AK004256.1 | - | 4 | 905 | ||
| AK159116.1 | - | 5 | 1020 | |||
| AK169537.1 | - | 4 | 934 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inip| Gene Symbol: Inip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013577.1 | NP_001013595.1 | 5 | 1032 | ||
| GenBank mRNA | AK004256.1 | - | 4 | 905 | ||
| AK159116.1 | - | 5 | 1020 | |||
| AK169537.1 | - | 4 | 934 | |||