siRNA Details
Gene Symbol: TTC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017931.2 | NP_060401.2 | 6 | 634 | ||
XM_011530259.1 | XP_011528561.1 | 7 | 762 | |||
XM_011530260.2 | XP_011528562.1 | 6 | 634 | |||
GenBank mRNA | AK000706.1 | - | 6 | 590 | ||
AK300606.1 | - | 5 | 440 | |||
BC018918.2 | - | 6 | 575 | 4130906 |
