siRNA Details
| Gene Symbol: Bod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024919.1 | NP_001020090.1 | 4 | 927 | ||
| GenBank mRNA | AK009485.1 | - | 4 | 909 | ||
| BC116435.1 | - | 5 | 759 | 40104481 | ||
| BC116436.1 | - | 5 | 759 | 40104482 | ||
| BC128508.1 | - | 5 | 829 | |||
| BC128509.1 | - | 5 | 759 | 40104485 | ||

Bod1
Genomic Map