siRNA Details
| Gene Symbol: Prm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013637.4 | NP_038665.1 | 2 | 317 | ||
| GenBank mRNA | AK006377.1 | - | 2 | 316 | ||
| AK007068.1 | - | 2 | 317 | |||
| AK018980.1 | - | 2 | 320 | |||
| BC049599.1 | - | 316 | 6773822 | |||
| BC059733.1 | - | 2 | 316 | 6774637 | ||
| K02926.1 | - | 2 | 302 | |||
| M27500.1 | - | 2 | 302 | |||
| X14003.1 | - | 2 | 316 | |||

Prm1
Genomic Map