siRNA Details
| Gene Symbol: PCDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105243.1 | NP_001098713.1 | 5 | 9043 | ||
| NM_001184880.1 | NP_001171809.1 | 6 | 9184 | |||
| NM_020766.2 | NP_065817.2 | 5 | 9040 | |||
| XM_011530997.2 | XP_011529299.1 | 6 | 8150 | |||
| GenBank mRNA | AB037734.1 | - | 5 | 5248 | ||
| AK055951.1 | - | 1 | 1589 | |||
| CR749278.1 | - | 5 | 5921 | |||
| EF676096.1 | - | 6 | 9184 | |||

PCDH19
Genomic Map