siRNA Details
| Gene Symbol: FAM234B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020853.1 | NP_065904.1 | 12, 13 | 1883 | ||
| XM_017019706.1 | XP_016875195.1 | 14, 15 | 2194 | |||
| GenBank mRNA | AB040900.1 | - | 9, 10 | 1290 | ||
| AK125278.1 | - | 12, 13 | 1883 | |||
| AK294732.1 | - | 6, 7 | 895 | |||
| BC039267.1 | - | 12, 13 | 1872 | 5274878 | ||
| BC132772.1 | - | 12, 13 | 1980 | 40146794 | ||
| BC132774.1 | - | 12, 13 | 1980 | 40146796 | ||
| CR936647.1 | - | 12, 13 | 1874 | |||

FAM234B
Genomic Map