siRNA Details
| Gene Symbol: NDUFV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001503.1 | NP_001001503.1 | 3 | 316 | ||
| NM_021075.3 | NP_066553.3 | 4 | 1411 | |||
| XM_011529586.2 | XP_011527888.1 | 5 | 1471 | |||
| XM_017028359.1 | XP_016883848.1 | 4 | 374 | |||
| GenBank mRNA | AK056041.1 | - | 6 | 1868 | ||
| AK289586.1 | - | 3 | 270 | |||
| AK308228.1 | - | 4 | 1368 | |||
| AY173949.1 | - | 3 | 274 | |||
| BC021217.2 | - | 4 | 1398 | 4302265 | ||
| BC033766.1 | - | 3 | 274 | |||
| BC054016.1 | - | 3 | 317 | |||
| CR542174.1 | - | 3 | 250 | |||
| CR542190.1 | - | 3 | 250 | |||

NDUFV3
Genomic Map