siRNA Details
| Gene Symbol: NDUFV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001503.1 | NP_001001503.1 | 3 | 984 | ||
| NM_021075.3 | NP_066553.3 | 4 | 2079 | |||
| XM_011529586.2 | XP_011527888.1 | 5 | 2139 | |||
| XM_017028359.1 | XP_016883848.1 | 4 | 1042 | |||
| GenBank mRNA | AY173949.1 | - | 3 | 935 | ||
| BC033766.1 | - | 3 | 943 | |||
| CF123536.1 | - | 1 | 599 | |||

NDUFV3
Genomic Map