siRNA Details
| Gene Symbol: Huwe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021523.4 | NP_067498.4 | 15 | 1797 | ||
| GenBank mRNA | AK031374.1 | - | 16 | 1824 | ||
| AK049460.1 | - | 15 | 1753 | |||
| AK142956.1 | - | 16 | 1784 | |||
| AK163416.1 | - | 16 | 1947 | |||
| AK165140.1 | - | 15 | 1733 | |||
| AK165648.1 | - | 16 | 1870 | |||
| AY772010.3 | - | 15 | 1734 | |||
| AY929611.1 | - | 13 | 1344 | |||
| DQ097265.1 | - | 15 | 1753 | |||

Huwe1
Genomic Map