siRNA Details
| Gene Symbol: Huwe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021523.4 | NP_067498.4 | 23 | 2738 | ||
| GenBank mRNA | AK031374.1 | - | 24 | 2765 | ||
| AK049460.1 | - | 23 | 2694 | |||
| AK142956.1 | - | 25 | 2962 | |||
| AK163416.1 | - | 24 | 2888 | |||
| AK165140.1 | - | 24 | 2908 | |||
| AY772010.3 | - | 23 | 2675 | |||
| AY929611.1 | - | 21 | 2285 | |||
| DQ097265.1 | - | 23 | 2694 | |||

Huwe1
Genomic Map