siRNA Details
| Gene Symbol: Ccnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022267.1 | NP_071603.1 | 5 | 1026 | ||
| GenBank mRNA | L09752.1 | - | 5 | 1026 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnd2| Gene Symbol: Ccnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022267.1 | NP_071603.1 | 5 | 1026 | ||
| GenBank mRNA | L09752.1 | - | 5 | 1026 | ||