siRNA Details
| Gene Symbol: 2810474O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 1929 | ||
| NM_001289662.1 | NP_001276591.1 | 3 | 1842 | |||
| NM_026054.3 | NP_080330.1 | 3 | 1860 | |||
| GenBank mRNA | AK031526.1 | - | 3 | 1725 | ||
| AK031989.1 | - | 4 | 1892 | |||
| AK079405.1 | - | 4 | 1760 | |||
| AK220403.1 | - | 3 | 1843 | |||
| BC086679.1 | - | 1 | 1282 | |||
| BC139435.1 | - | 3 | 1782 | 8862457 | ||
| BC139436.1 | - | 4 | 1869 | 8862458 | ||

2810474O19Rik
Genomic Map