siRNA Details
| Gene Symbol: Plxnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026376.3 | NP_080652.2 | 7 | 2322 | ||
| GenBank mRNA | AK129175.1 | - | 7 | 1587 | ||
| AK147513.1 | - | 7 | 2323 | |||
| AK155833.1 | - | 5 | 545 | |||
| AY688678.1 | - | 7 | 2282 | |||

Plxnd1
Genomic Map