siRNA Details
| Gene Symbol: PTCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017952.5 | NP_060422.4 | 15, 16 | 1298 | ||
| GenBank mRNA | AK315676.1 | - | 9, 10 | 752 | ||
| AL832520.1 | - | 4, 5 | 302 | |||
| AY277598.1 | - | 3, 4 | 266 | |||
| BC001758.1 | - | 4, 5 | 372 | |||
| BC011832.2 | - | 15, 16 | 1233 | 3939273 | ||

PTCD3
Genomic Map