siRNA Details
| Gene Symbol: Ngdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026890.2 | NP_081166.1 | 7 | 627 | ||
| GenBank mRNA | AK005098.1 | - | 7 | 509 | ||
| AK200275.1 | - | 5 | 377 | |||
| AK212411.1 | - | 5 | 377 | |||
| AK214945.1 | - | 5 | 377 | |||
| BC023770.1 | - | 6 | 512 | |||
| BC052790.1 | - | 7 | 565 | 1515292 | ||

Ngdn
Genomic Map