siRNA Details
| Gene Symbol: Wdr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185162.1 | NP_001172091.1 | 3 | 570 | ||
| NM_026898.2 | NP_081174.1 | 3 | 802 | |||
| GenBank mRNA | AK005105.1 | - | 3 | 769 | ||
| BC028850.1 | - | 2 | 534 | 3987279 | ||

Wdr53
Genomic Map