siRNA Details
| Gene Symbol: Ptms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026988.2 | NP_081264.1 | 5 | 933 | ||
| GenBank mRNA | AK011360.1 | - | 5 | 933 | ||
| BC052699.1 | - | 5 | 918 | 5715702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptms| Gene Symbol: Ptms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026988.2 | NP_081264.1 | 5 | 933 | ||
| GenBank mRNA | AK011360.1 | - | 5 | 933 | ||
| BC052699.1 | - | 5 | 918 | 5715702 | ||