siRNA Details
| Gene Symbol: Pacrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027032.2 | NP_081308.1 | 4 | 836 | ||
| GenBank mRNA | AK005771.1 | - | 4 | 832 | ||
| AK135510.1 | - | 4 | 838 | |||
| BC120740.1 | - | 4 | 814 | 40129663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pacrg| Gene Symbol: Pacrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027032.2 | NP_081308.1 | 4 | 836 | ||
| GenBank mRNA | AK005771.1 | - | 4 | 832 | ||
| AK135510.1 | - | 4 | 838 | |||
| BC120740.1 | - | 4 | 814 | 40129663 | ||