siRNA Details
Gene Symbol: 1700030J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027103.2 | NP_081379.1 | 3 | 2904 | ||
GenBank mRNA | AK030075.1 | - | 3 | 2905 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700030J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027103.2 | NP_081379.1 | 3 | 2904 | ||
GenBank mRNA | AK030075.1 | - | 3 | 2905 |