siRNA Details
| Gene Symbol: 1700030J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027103.2 | NP_081379.1 | 3 | 714 | ||
| GenBank mRNA | AK006553.1 | - | 3 | 719 | ||
| AK030075.1 | - | 3 | 715 | |||
| AK031529.1 | - | 3 | 599 | |||
| BC049630.1 | - | 3 | 643 | |||
| BC100501.1 | - | 3 | 734 | 30917058 | ||
| BC137750.1 | - | 3 | 652 | 8860769 | ||
| BC137751.1 | - | 3 | 652 | 8860770 | ||

1700030J22Rik
Genomic Map