siRNA Details
| Gene Symbol: EPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017957.2 | NP_060427.2 | 10 | 2924 | ||
| GenBank mRNA | AK000785.1 | - | 10 | 2873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPN3| Gene Symbol: EPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017957.2 | NP_060427.2 | 10 | 2924 | ||
| GenBank mRNA | AK000785.1 | - | 10 | 2873 | ||