siRNA Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027238.2 | NP_081514.1 | 3 | 239 | ||
| GenBank mRNA | AK007856.1 | - | 3 | 240 | ||
| AK037208.1 | - | 3 | 240 | |||
| AK049108.1 | - | 3 | 290 | |||
| BC151021.1 | - | 3 | 201 | 9087934 | ||

Ttc39b
Genomic Map