siRNA Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027238.2 | NP_081514.1 | 10 | 839 | ||
| GenBank mRNA | AK037208.1 | - | 10 | 840 | ||
| AK049108.1 | - | 10 | 890 | |||
| AK132655.1 | - | 5 | 290 | |||
| BC151021.1 | - | 10 | 801 | 9087934 | ||

Ttc39b
Genomic Map