siRNA Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027295.2 | NP_081571.1 | 7 | 1158 | ||
| GenBank mRNA | AK159493.1 | - | 7 | 1159 | ||
| AK211503.1 | - | 1 | 128 | |||
| AK213358.1 | - | 1 | 128 | |||
| BC004580.1 | - | 7 | 1158 | 3587008 | ||

Rab28
Genomic Map