siRNA Details
| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027426.3 | NP_081702.2 | 7 | 2166 | ||
| GenBank mRNA | AK086198.1 | - | 7 | 2167 | ||
| BC057066.1 | - | 1 | 36 | |||
| BC058734.1 | - | 3 | 1282 | |||
| BC060239.1 | - | 7 | 2147 | 6853328 | ||

Draxin
Genomic Map