siRNA Details
| Gene Symbol: Pnma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027438.3 | NP_081714.2 | 1 | 1146 | ||
| GenBank mRNA | AK017476.1 | - | 1 | 940 | ||
| AK028331.1 | - | 1 | 1084 | |||
| AK136933.1 | - | 1 | 1148 | |||
| BC116687.1 | - | 1 | 900 | 40106117 | ||
| BC116688.1 | - | 1 | 896 | 40106116 | ||

Pnma1
Genomic Map