siRNA Details
| Gene Symbol: 3830417A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027512.2 | NP_081788.2 | 4 | 1106 | ||
| GenBank mRNA | AK014439.1 | - | 4 | 1103 | ||
| AK049539.1 | - | 4 | 1107 | |||
| BC068255.1 | - | 4 | 1106 | 30021331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3830417A13Rik| Gene Symbol: 3830417A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027512.2 | NP_081788.2 | 4 | 1106 | ||
| GenBank mRNA | AK014439.1 | - | 4 | 1103 | ||
| AK049539.1 | - | 4 | 1107 | |||
| BC068255.1 | - | 4 | 1106 | 30021331 | ||