siRNA Details
| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027724.2 | NP_082000.2 | 5 | 1335 | ||
| GenBank mRNA | AY695827.1 | - | 4 | 1070 | ||
| BC082568.1 | - | 4 | 1237 | 30549853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cage1| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027724.2 | NP_082000.2 | 5 | 1335 | ||
| GenBank mRNA | AY695827.1 | - | 4 | 1070 | ||
| BC082568.1 | - | 4 | 1237 | 30549853 | ||