siRNA Details
| Gene Symbol: Pex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293806.1 | NP_001280735.1 | 9 | 1933 | ||
| NM_027777.2 | NP_082053.1 | 8 | 1813 | |||
| GenBank mRNA | AK082305.1 | - | 2 | 107 | ||
| AK087687.1 | - | 4 | 371 | |||
| BC090845.1 | - | 8 | 1813 | 6400629 | ||
| DV646280.1 | - | 5 | 574 | |||

Pex1
Genomic Map