siRNA Details
| Gene Symbol: IWS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017969.2 | NP_060439.2 | 3 | 498 | ||
| XM_005263707.3 | XP_005263764.1 | 3 | 334 | |||
| XM_006712626.3 | XP_006712689.1 | 3 | 412 | |||
| XM_011511450.1 | XP_011509752.1 | 3 | 412 | |||
| XM_011511452.1 | XP_011509754.1 | 3 | 412 | |||
| XM_017004467.1 | XP_016859956.1 | 3 | 499 | |||
| XM_017004468.1 | XP_016859957.1 | 3 | 268 | |||
| GenBank mRNA | AK027561.1 | - | 3 | 455 | ||
| AK294669.1 | - | 3 | 476 | |||
| AK295970.1 | - | 3 | 261 | |||
| AK296848.1 | - | 3 | 450 | |||
| AK309006.1 | - | 2 | 353 | |||
| AL834178.1 | - | 3 | 446 | |||
| BC065279.1 | - | 3 | 257 | |||
| BC110536.1 | - | 3 | 328 | 40035173 | ||
| BC110537.1 | - | 3 | 328 | 40035174 | ||
| DA268882.1 | - | 3 | 499 | |||

IWS1
Genomic Map